Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGCGTTTGGCGAAAGGGAAACC[A/T]TCTTTCTCCTTCCGAAGTGCCCCGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ltv1 PubMed Links |
Ltv1 - LTV1 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181470.4 | Intron | NP_852135.1 |