Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCTGTCACCTGGGCCTGGAAC[A/G]TTCTGCCCTTGATTTCTCAGGCTGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kif5c PubMed Links |
Kif5c - kinesin family member 5C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008449.2 | Intron | NP_032475.2 |