Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAGCAGGTGTCTTAAAACCCCAG[C/T]GTCAAAAACTGATCAGTGATAGTGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ncald PubMed Links |
Ncald - neurocalcin delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170866.1 | Intron | NP_001164337.1 | ||||
NM_001170867.1 | Intron | NP_001164338.1 | ||||
NM_001170868.1 | Intron | NP_001164339.1 | ||||
NM_134094.4 | Intron | NP_598855.2 |