Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bmyc PubMed Links |
Bmyc - brain expressed myelocytomatosis oncogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Obp2b - odorant binding protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099301.1 | Intron | NP_001092771.1 |