Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTTCGGCTTCTCCCAGCATTAG[A/G]CTTGTGTTCCTCACCCGCCCAAGTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Syne2 PubMed Links |
Syne2 - spectrin repeat containing, nuclear envelope 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005510.2 | Intron | NP_001005510.2 |