Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCCAAGTATTGGTGTCTTTTA[G/T]ATCAGTCTGAGTTTTGAATTGATGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bank1 PubMed Links |
Bank1 - B cell scaffold protein with ankyrin repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033350.2 | Intron | NP_001028522.2 |