Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAAAGAGAAATTATCCAGGGCAC[A/G]GAGGAGGCAACCCTGGTTCAACTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ano4 PubMed Links |
Ano4 - anoctamin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277188.1 | Intron | NP_001264117.1 |