Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCACCCTGGGCATCTGGAGGGAC[A/G]TCTGAATGATAAGAAGAACTGTCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
B4galt1 PubMed Links |
B4galt1 - UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022305.4 | 2057 | UTR 3 | NP_071641.1 |