Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTATGGATTCAACCTATAGCATT[G/T]CTATGGCATGGATTGATAGTTTCAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mgat4c PubMed Links |
Mgat4c - MGAT4 family, member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162368.1 | Intron | NP_001155840.1 | ||||
NM_001162369.1 | Intron | NP_001155841.1 | ||||
NM_001205098.1 | Intron | NP_001192027.1 | ||||
NM_026243.4 | Intron | NP_080519.2 |