Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTATCATGAGGTGAGTGGGCTCC[A/G]TTTCTCACTACAATGCCATCTTCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sash1 PubMed Links |
Sash1 - SAM and SH3 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175155.4 | Intron | NP_780364.3 |