Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCATTCCTTTCCGGCCTTTCAT[C/G]GGGAAGTTGATGTCGGTTAGAACAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Adgrb3 PubMed Links |
Adgrb3 - adhesion G protein-coupled receptor B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175642.4 | 2424 | Silent Mutation | NP_783573.4 |