Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ccr10 PubMed Links |
Ccr10 - chemokine (C-C motif) receptor 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Cntnap1 - contactin associated protein-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Plekhh3 - pleckstrin homology domain containing, family H (with MyTH4 domain) member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_146030.2 | 1721 | Silent Mutation | NP_666142.1 |
Tubg2 - tubulin, gamma 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |