Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATCAATAAAGAATATATCTATGG[A/T]AATCTGGGATATGAACTTATGTAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tmem67 PubMed Links |
Tmem67 - transmembrane protein 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177861.4 | 1908 | Missense Mutation | NP_808529.2 |