Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGAAATTATCTTTCTTAGGCACA[C/T]TGAAGGACCTTTGTAACAAAGGAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Clvs1 PubMed Links |
Clvs1 - clavesin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028940.2 | Intron | NP_083216.1 |