Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Eml6 PubMed Links |
Eml6 - echinoderm microtubule associated protein like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_146016.2 | 6089 | UTR 3 | NP_666128.2 |
Rtn4 - reticulon 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |