Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATACCTCAAAGATTTCAAACCC[A/G]GCAATATCCAGAATGCCCAGGAAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Myh11 PubMed Links |
Myh11 - myosin, heavy polypeptide 11, smooth muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161775.1 | 1532 | Silent Mutation | NP_001155247.1 | |||
NM_013607.2 | 1532 | Silent Mutation | NP_038635.2 |