Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTCAGCCCCAGGCCCTTCCCAC[C/A]CATGCCGTGTGGCCCTGGGCAGGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sspo PubMed Links |
Sspo - SCO-spondin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173428.3 | 7154 | Missense Mutation | NP_775604.3 |