Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Spink14 PubMed Links |
Spink14 - serine peptidase inhibitor, Kazal type 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Spink5 - serine peptidase inhibitor, Kazal type 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081180.1 | 4269 | UTR 3 | NP_001074649.1 |