Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rpl38 PubMed Links |
Rpl38 - ribosomal protein L38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048057.1 | Intron | NP_001041522.1 | ||||
NM_001048058.1 | Intron | NP_001041523.1 | ||||
NM_023372.2 | Intron | NP_075861.1 |
Ttyh2 - tweety homolog 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |