Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCCTTATAAAACACAAAGGCAC[A/G]ACACTCATTGTCAGATCAACCAAGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rps6kc1 PubMed Links |
Rps6kc1 - ribosomal protein S6 kinase polypeptide 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178775.4 | Intron | NP_848890.3 |