Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCTTCTGGTTTCTAAAGCCTCA[A/C]ATTGTATGAGCTGAGTCTTCCTGTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Daam1 PubMed Links |
Daam1 - dishevelled associated activator of morphogenesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286452.1 | Intron | NP_001273381.1 | ||||
NM_026102.3 | Intron | NP_080378.2 | ||||
NM_172464.3 | Intron | NP_766052.2 |