Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCGAACAAAAACAAATGAGGATC[T/G]GTCAAAGTCGTACCACTGCTTTGCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hectd1 PubMed Links |
Hectd1 - HECT domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144788.2 | 3498 | Silent Mutation | NP_659037.2 |