Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACTAGAGGAGCTCTTCCCCACA[C/T]CTGGGGAGAAGCAAGTCACCCTCTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lrp1b PubMed Links |
Lrp1b - low density lipoprotein-related protein 1B (deleted in tumors) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053011.2 | Intron | NP_443737.2 |