Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGACCAGGTCTACACAGCCATGC[C/T]CTGAAGATGCCTGTCTTCACAGAAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ints9 PubMed Links |
Ints9 - integrator complex subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253731.1 | Intron | NP_001240660.1 | ||||
NM_153414.4 | Intron | NP_700463.2 |