Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGAGAACACCTTCATTTCCAGAT[C/T]ACTAATGCGTCTGCTTTTATAAAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Aldh1a1 PubMed Links |
Aldh1a1 - aldehyde dehydrogenase family 1, subfamily A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013467.3 | Intron | NP_038495.2 |