Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATACAGCTACGATGTAAGATTTT[C/G]TTGGCTAAAGATTTCCCAGCATAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sptlc3 PubMed Links |
Sptlc3 - serine palmitoyltransferase, long chain base subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175467.3 | Intron | NP_780676.1 |