Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTTCACGCTGTTGTCTCTCAAA[C/T]TCAAGTCTAATGAGATCAAATGGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kif16b PubMed Links |
Kif16b - kinesin family member 16B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081133.2 | 1884 | Silent Mutation | NP_001074602.1 |