Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAGTCACCATGGACCAACTGCC[A/G]ATGATTTCTGGATATAGTGGACAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc25a21 PubMed Links |
Slc25a21 - solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167976.1 | Intron | NP_001161448.1 | ||||
NM_172577.3 | Intron | NP_766165.2 |