Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGTAAGCTCCTGATTCGAAGCT[C/G]GTCTGACATCCGGTGCAAGTGATGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kif13a PubMed Links |
Kif13a - kinesin family member 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010617.2 | 4890 | Missense Mutation | NP_034747.2 |