Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATATTTATGGGAATTGTTGTTC[C/T]CACTATGAAGGCATCTGTTATTGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Vit PubMed Links |
Vit - vitrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197028.1 | 298 | Missense Mutation | NP_001183957.1 | |||
NM_028813.2 | 298 | Missense Mutation | NP_083089.1 |