Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCAACCAATAAGCGGATTTAGG[C/T]ACTGAGTGCAGTCGTGGAGTTTATT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnt2 PubMed Links |
Kcnt2 - potassium channel, subfamily T, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081027.2 | Intron | NP_001074496.2 |