Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGGAAGTAAGCGATACAGTAAT[A/G]TCAATGCATTATAATGGACACAGGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dst PubMed Links |
Dst - dystonin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276764.1 | Intron | NP_001263693.1 | ||||
NM_010081.2 | Intron | NP_034211.2 | ||||
NM_133833.3 | Intron | NP_598594.2 | ||||
NM_134448.3 | Intron | NP_604443.2 |