Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGACAGAGCTGGCTAGATTCACC[G/A]AGGAGCTGGAATGGTGGTGTCTGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fgf12 PubMed Links |
Fgf12 - fibroblast growth factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276420.1 | Intron | NP_001263349.1 |