Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTATGCTTTTGCTGATCCCCA[A/G]CCATAAAATTATTTTTGTTTCTACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
5830433I10Rik PubMed Links |
5830433I10Rik - RIKEN cDNA 5830433I10 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Chd1 - chromodomain helicase DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007690.3 | Intron | NP_031716.2 |