Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGTCACGCACCTCCACGTCACAG[C/T]GCTAGAGCTGCGATACTTTTCCTAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc35b3 PubMed Links |
Slc35b3 - solute carrier family 35, member B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170430.1 | Intron | NP_001163901.1 | ||||
NM_001170431.1 | Intron | NP_001163902.1 | ||||
NM_134060.4 | Intron | NP_598821.1 |