Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTAGACTGCTCAGTTGTACTGCC[A/T]GTATCGCATGATTCTAATCCTAAGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lep PubMed Links |
Lep - leptin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008493.3 | Intron | NP_032519.1 |