Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGGCCTTCTGGTATCAGACAA[C/T]TTATTTACAAAGGGTGATCATTCAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cpsf6 PubMed Links |
Cpsf6 - cleavage and polyadenylation specific factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013391.2 | 6185 | UTR 3 | NP_001013409.1 |