Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGATTGGGAAAATCACGTAAAAG[C/T]GCACCAATTAAAGACACTACAAGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Apaf1 PubMed Links |
Apaf1 - apoptotic peptidase activating factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042558.1 | 1655 | Silent Mutation | NP_001036023.1 | |||
NM_001282947.1 | 1655 | Silent Mutation | NP_001269876.1 | |||
NM_009684.2 | 1655 | Silent Mutation | NP_033814.2 |