Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACGGTAAGGACCCTGGAGAGGGT[C/T]TGGGCTCATAGCCAGCAGGTGTGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnk2 PubMed Links |
Kcnk2 - potassium channel, subfamily K, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159850.1 | Intron | NP_001153322.1 | ||||
NM_001281847.1 | Intron | NP_001268776.1 | ||||
NM_001281848.1 | Intron | NP_001268777.1 | ||||
NM_010607.3 | Intron | NP_034737.2 |