Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGTGGGGGGATTAGGTCAACTG[C/T]AAGTACTTGTATTGTTTCACTTTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tmtc2 PubMed Links |
Tmtc2 - transmembrane and tetratricopeptide repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177368.4 | Intron | NP_796342.2 |