Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTTACCTGACTGGAGATGGGGT[C/T]GGTGCCCGCACGTGTCTGACTGGGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ttn PubMed Links |
Ttn - titin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011652.3 | 1117 | Silent Mutation | NP_035782.3 | |||
NM_028004.2 | 1117 | Silent Mutation | NP_082280.2 |