Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACTCCACCTTCCTTCCTGGTTC[A/G]ATGCCAAATGATGCCGTAACAACCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Acss3 PubMed Links |
Acss3 - acyl-CoA synthetase short-chain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142804.1 | 744 | Silent Mutation | NP_001136276.1 | |||
NM_198636.3 | 744 | Silent Mutation | NP_941038.2 |