Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGGGAGGACATATTGAATATA[A/G]AACCCATAACATCACATTTCCAGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
A330070K13Rik PubMed Links |
A330070K13Rik - RIKEN cDNA A330070K13 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Caln1 - calneuron 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021371.2 | Intron | NP_067346.1 | ||||
NM_181045.1 | Intron | NP_851388.1 |