Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCACACTACATTGGTACAGGGC[A/G]GAACCTTTTTTAGGCGACATGCTAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cadps PubMed Links |
Cadps - Ca2+-dependent secretion activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042617.1 | Intron | NP_001036082.1 | ||||
NM_012061.3 | Intron | NP_036191.2 |