Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAGACTCAGAGGGTCACAGGTC[C/T]CTGCTGCAACTCCAAAAACAGAGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dnajc10 PubMed Links |
Dnajc10 - DnaJ (Hsp40) homolog, subfamily C, member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024181.2 | 3942 | UTR 3 | NP_077143.2 |