Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm40266 PubMed Links |
Gm40266 - predicted gene, 40266 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Sema3a - sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243072.1 | Intron | NP_001230001.1 | ||||
NM_001243073.1 | Intron | NP_001230002.1 | ||||
NM_009152.4 | Intron | NP_033178.2 |