Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTGATCACTTCCTTCTGTGCTA[C/T]AGTCAACCTCTGGATTTCCTCCAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Usp9y PubMed Links |
Usp9y - ubiquitin specific peptidase 9, Y chromosome | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_148943.2 | Intron | NP_683745.2 |