Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGATCATACAGACTCCCCCCTTT[G/T]GTCAACTCTGGTTAAATACTGAATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Smurf2 PubMed Links |
Smurf2 - SMAD specific E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025481.2 | Intron | NP_079757.2 |