Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cage1 PubMed Links |
Cage1 - cancer antigen 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Ssr1 - signal sequence receptor, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025965.3 | 608 | Silent Mutation | NP_080241.3 |