Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ngly1 PubMed Links |
Ngly1 - N-glycanase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Oxsm - 3-oxoacyl-ACP synthase, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027695.3 | 1161 | Silent Mutation | NP_081971.1 |